Assay Details
| Gene Symbol: Hsepi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136405.4 | NP_610249.2 | 1 - 2 | 430 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsepi| Gene Symbol: Hsepi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136405.4 | NP_610249.2 | 1 - 2 | 430 | 77 | |
Set Membership: |
|