Assay Details
| Gene Symbol: mRpL52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136469.4 | NP_610313.1 | 2 - 3 | 293 | 70 | |
| GenBank mRNA | AY113553.1 | - | 2 - 3 | 297 | 70 | |
| KX531389.1 | - | 2 - 3 | 196 | 70 | ||
More Information
Set Membership: |
|

mRpL52
Genomic Map