Assay Details
Gene Symbol: Vha44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144207.2 | NP_001137679.1 | 3 - 4 | 449 | 98 | |
NM_166163.3 | NP_725564.1 | 3 - 4 | 504 | 98 | ||
NM_166164.5 | NP_725565.3 | 3 - 4 | 504 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
