Assay Details
Gene Symbol: Vha44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144207.2 | NP_001137679.1 | 6 - 7 | 1975 | 83 | |
NM_057918.5 | NP_477266.1 | 4 - 5 | 631 | 83 | ||
NM_134313.4 | NP_599140.1 | 4 - 5 | 686 | 83 | ||
NM_166163.3 | NP_725564.1 | 5 - 6 | 848 | 83 | ||
NM_166164.5 | NP_725565.3 | 6 - 7 | 2030 | 83 | ||
GenBank mRNA | AY061038.1 | - | 4 - 5 | 665 | 83 | |
AY102676.1 | - | 4 - 5 | 631 | 83 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
