Assay Details
Gene Symbol: Vha44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144207.2 | NP_001137679.1 | 10 - 11 | 2570 | 69 | |
NM_057918.5 | NP_477266.1 | 8 - 9 | 1226 | 69 | ||
NM_134313.4 | NP_599140.1 | 8 - 9 | 1281 | 69 | ||
NM_166163.3 | NP_725564.1 | 9 - 10 | 1443 | 69 | ||
NM_166164.5 | NP_725565.3 | 10 - 11 | 2625 | 69 | ||
GenBank mRNA | AY061038.1 | - | 8 - 9 | 1260 | 69 | |
AY102676.1 | - | 8 - 9 | 1226 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
