Assay Details
| Gene Symbol: Hmgs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299559.1 | NP_001286488.1 | 2 | 1746 | 113 | |
| NM_079972.4 | NP_524711.1 | 2 | 2416 | 113 | ||
| NM_166166.3 | NP_725567.1 | 2 | 1762 | 113 | ||
| NM_166167.3 | NP_725568.1 | 2 | 1777 | 113 | ||
| NM_166168.3 | NP_725569.1 | 2 | 1725 | 113 | ||
| NM_166169.3 | NP_725570.1 | 1 | 1649 | 113 | ||
| GenBank mRNA | AY051743.1 | - | 2 | 2416 | 113 | |
More Information
Set Membership: |
|

Hmgs
Genomic Map