Assay Details
Gene Symbol: Sply | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143775.3 | NP_652032.1 | 3 - 4 | 974 | 92 | |
NM_166215.2 | NP_725652.1 | 2 - 3 | 911 | 92 | ||
GenBank mRNA | AJ297394.1 | - | 2 - 3 | 890 | 92 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
