Assay Details
Gene Symbol: Prosalpha5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057854.4 | NP_477202.2 | 1 - 2 | 200 | 59 | |
NM_166221.4 | NP_725669.1 | 2 - 3 | 211 | 59 | ||
GenBank mRNA | AY061404.1 | - | 2 - 3 | 195 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
