Assay Details
| Gene Symbol: eIF3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137384.3 | NP_611228.1 | 2 - 3 | 244 | 56 | |
| NM_166239.4 | NP_725691.1 | 1 - 2 | 249 | 56 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eIF3b| Gene Symbol: eIF3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137384.3 | NP_611228.1 | 2 - 3 | 244 | 56 | |
| NM_166239.4 | NP_725691.1 | 1 - 2 | 249 | 56 | ||
Set Membership: |
|