Assay Details
Gene Symbol: grh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259472.1 | NP_001246401.1 | 11 - 12 | 2052 | 79 | |
NM_001299622.1 | NP_001286551.1 | 11 - 12 | 5103 | 79 | ||
NM_057494.4 | NP_476842.2 | 10 - 11 | 3641 | 79 | ||
NM_057495.4 | NP_476843.2 | 11 - 12 | 3548 | 79 | ||
NM_057496.5 | NP_476844.2 | 13 - 14 | 4358 | 79 | ||
NM_057497.4 | NP_476845.2 | 12 - 13 | 5072 | 79 | ||
NM_166250.4 | NP_725723.2 | 10 - 11 | 2311 | 79 | ||
NM_166252.3 | NP_725725.1 | 10 - 11 | 2139 | 79 | ||
GenBank mRNA | BT029431.1 | - | 11 - 12 | 3242 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
