Assay Details
Gene Symbol: grh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259472.1 | NP_001246401.1 | 6 - 7 | 1502 | 72 | |
NM_001299622.1 | NP_001286551.1 | 7 - 8 | 4460 | 72 | ||
NM_057494.4 | NP_476842.2 | 6 - 7 | 2998 | 72 | ||
NM_057495.4 | NP_476843.2 | 6 - 7 | 2998 | 72 | ||
NM_057496.5 | NP_476844.2 | 8 - 9 | 3808 | 72 | ||
NM_057497.4 | NP_476845.2 | 8 - 9 | 4429 | 72 | ||
NM_166250.4 | NP_725723.2 | 6 - 7 | 1668 | 72 | ||
NM_166252.3 | NP_725725.1 | 6 - 7 | 1496 | 72 | ||
GenBank mRNA | BT029431.1 | - | 6 - 7 | 2692 | 72 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
