Assay Details
Gene Symbol: Rgk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137498.3 | NP_611342.1 | 4 - 5 | 2402 | 66 | |
GenBank mRNA | BT011477.1 | - | 4 - 5 | 2402 | 66 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
