Assay Details
Gene Symbol: Rgk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299648.1 | NP_001286577.1 | 5 - 6 | 2299 | 81 | |
NM_137498.3 | NP_611342.1 | 5 - 6 | 2680 | 81 | ||
GenBank mRNA | BT011477.1 | - | 5 - 6 | 2680 | 81 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
