Assay Details
Gene Symbol: Rgk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299648.1 | NP_001286577.1 | 6 - 7 | 2402 | 89 | |
NM_137498.3 | NP_611342.1 | 6 - 7 | 2783 | 89 | ||
GenBank mRNA | BT011477.1 | - | 6 - 7 | 2783 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
