Assay Details
| Gene Symbol: 5-HT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001369983.1 | NP_001356890.1 | 8 - 9 | 2585 | 80 | |
| NM_166322.2 | NP_725849.1 | 8 - 9 | 2585 | 80 | ||
More Information
Set Membership: |
|

5-HT1A
Genomic Map