Assay Details
Gene Symbol: Rgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166341.3 | NP_725875.3 | 4 - 5 | 1356 | 67 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
