Assay Details
Gene Symbol: Fak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144246.3 | NP_001137718.1 | 10 - 11 | 3014 | 68 | |
NM_001144247.3 | NP_001137719.2 | 11 - 12 | 2443 | 68 | ||
NM_001169736.2 | NP_001163207.1 | 10 - 11 | 3014 | 68 | ||
NM_079069.4 | NP_523793.2 | 10 - 11 | 3014 | 68 | ||
NM_166352.3 | NP_725891.1 | 11 - 12 | 2800 | 68 | ||
NM_166353.3 | NP_725892.1 | 11 - 12 | 2541 | 68 | ||
GenBank mRNA | BT133413.1 | - | 10 - 11 | 2997 | 68 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
