Assay Details
Gene Symbol: Fak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144247.3 | NP_001137719.2 | 12 - 13 | 2585 | 85 | |
NM_001169736.2 | NP_001163207.1 | 11 - 12 | 3156 | 85 | ||
NM_079069.4 | NP_523793.2 | 11 - 12 | 3156 | 85 | ||
NM_166352.3 | NP_725891.1 | 12 - 13 | 2942 | 85 | ||
NM_166353.3 | NP_725892.1 | 12 - 13 | 2683 | 85 | ||
GenBank mRNA | BT133413.1 | - | 11 - 12 | 3139 | 85 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
