Assay Details
| Gene Symbol: Fak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144246.3 | NP_001137718.1 | 2 - 3 | 1419 | 153 | |
| NM_001144247.3 | NP_001137719.2 | 3 - 4 | 848 | 153 | ||
| NM_001169736.2 | NP_001163207.1 | 2 - 3 | 1419 | 153 | ||
| NM_079069.4 | NP_523793.2 | 2 - 3 | 1419 | 153 | ||
| NM_166352.3 | NP_725891.1 | 3 - 4 | 1205 | 153 | ||
| NM_166353.3 | NP_725892.1 | 3 - 4 | 946 | 153 | ||
| GenBank mRNA | BT133413.1 | - | 2 - 3 | 1402 | 153 | |
More Information
Set Membership: |
|

Fak
Genomic Map