Assay Details
| Gene Symbol: EloC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079070.4 | NP_523794.1 | 1 - 2 | 357 | 66 | |
| NM_166354.3 | NP_725894.1 | 2 - 3 | 279 | 66 | ||
| GenBank mRNA | AY070956.1 | - | 2 - 3 | 269 | 66 | |
| AY113630.1 | - | 2 - 3 | 277 | 66 | ||
| KX531788.1 | - | 1 - 2 | 210 | 66 | ||
More Information
Set Membership: |
|

EloC
Genomic Map