Assay Details
| Gene Symbol: sm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032268.3 | NP_001027439.2 | 2 - 3 | 1055 | 78 | |
| NM_001032269.3 | NP_001027440.1 | 2 - 3 | 778 | 78 | ||
| NM_001259494.2 | NP_001246423.1 | 2 - 3 | 1055 | 78 | ||
| NM_001259496.2 | NP_001246425.1 | 2 - 3 | 1055 | 78 | ||
| NM_001259497.2 | NP_001246426.1 | 2 - 3 | 1055 | 78 | ||
| NM_001259498.2 | NP_001246427.1 | 2 - 3 | 778 | 78 | ||
| NM_001259499.2 | NP_001246428.1 | 2 - 3 | 1055 | 78 | ||
| NM_001259501.2 | NP_001246430.1 | 2 - 3 | 1055 | 78 | ||
| NM_001259504.2 | NP_001246433.1 | 2 - 3 | 778 | 78 | ||
| NM_001274164.1 | NP_001261093.1 | 2 - 3 | 740 | 78 | ||
| NM_057383.7 | NP_476731.1 | 2 - 3 | 1055 | 78 | ||
| GenBank mRNA | BT011352.1 | - | 2 - 3 | 1048 | 78 | |
| X97706.1 | - | 1049 | 78 | |||
More Information
Set Membership: |
|

sm
Genomic Map