Assay Details
| Gene Symbol: sm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032269.3 | NP_001027440.1 | 5 - 6 | 1166 | 57 | |
| NM_057383.7 | NP_476731.1 | 5 - 6 | 1443 | 57 | ||
| GenBank mRNA | X97706.1 | - | 1437 | 57 | ||
More Information
Set Membership: |
|

sm
Genomic Map