Assay Details
| Gene Symbol: Obp56f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166375.3 | NP_725926.1 | 1 - 2 | 77 | 86 | |
| GenBank mRNA | AF457146.1 | - | 1 - 2 | 53 | 86 | |
| BT023320.1 | - | 1 - 2 | 58 | 86 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  3' Most  Recommended Assay  Drosophila melanogaster | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Obp56f
Obp56f Genomic Map
Genomic Map Back To Top
 Back To Top