Assay Details
| Gene Symbol: Obp56i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166376.2 | NP_725929.1 | 1 - 2 | 91 | 80 | |
| GenBank mRNA | BT023325.1 | - | 1 - 2 | 91 | 80 | |
| BT029404.1 | - | 1 - 2 | 91 | 80 | ||
More Information
Set Membership: |
|

Obp56i
Genomic Map