Assay Details
| Gene Symbol: CG3216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137688.2 | NP_611532.2 | 4 - 5 | 656 | 63 | |
| NM_166415.2 | NP_726013.2 | 4 - 5 | 656 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG3216| Gene Symbol: CG3216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137688.2 | NP_611532.2 | 4 - 5 | 656 | 63 | |
| NM_166415.2 | NP_726013.2 | 4 - 5 | 656 | 63 | ||
Set Membership: |
|