Assay Details
| Gene Symbol: Pu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166430.3 | NP_726037.1 | 1 - 2 | 357 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pu| Gene Symbol: Pu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166430.3 | NP_726037.1 | 1 - 2 | 357 | 73 | |
Set Membership: |
|