Assay Details
Gene Symbol: Pu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166431.4 | NP_726038.1 | 1 - 2 | 525 | 60 | |
GenBank mRNA | AY051890.1 | - | 1 - 2 | 508 | 60 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
