Assay Details
Gene Symbol: Sdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144256.2 | NP_001137728.1 | 3 - 4 | 538 | 110 | |
NM_001144257.4 | NP_001137729.2 | 3 - 4 | 538 | 110 | ||
NM_001169767.3 | NP_001163238.2 | 3 - 4 | 538 | 110 | ||
NM_001259518.2 | NP_001246447.1 | 2 - 3 | 304 | 110 | ||
NM_001299749.1 | NP_001286678.1 | 2 - 3 | 304 | 110 | ||
NM_001369961.1 | NP_001356968.1 | 3 - 4 | 538 | 110 | ||
NM_057617.5 | NP_476965.1 | 3 - 4 | 538 | 110 | ||
NM_166449.5 | NP_726071.1 | 3 - 4 | 560 | 110 | ||
NM_166450.3 | NP_726072.2 | 3 - 4 | 538 | 110 | ||
GenBank mRNA | AY069377.1 | - | 3 - 4 | 531 | 110 | |
AY089610.1 | - | 3 - 4 | 535 | 110 | ||
BT125826.1 | - | 3 - 4 | 498 | 110 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001259518.2, NM_001299749.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
