Assay Details
| Gene Symbol: Sdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144256.2 | NP_001137728.1 | 6 - 7 | 1363 | 60 | |
| NM_001144257.4 | NP_001137729.2 | 6 - 7 | 1651 | 60 | ||
| NM_001169767.3 | NP_001163238.2 | 6 - 7 | 1654 | 60 | ||
| NM_001259518.2 | NP_001246447.1 | 4 - 5 | 613 | 60 | ||
| NM_001299749.1 | NP_001286678.1 | 5 - 6 | 1129 | 60 | ||
| NM_001369961.1 | NP_001356968.1 | 6 - 7 | 1366 | 60 | ||
| NM_057617.5 | NP_476965.1 | 6 - 7 | 1366 | 60 | ||
| NM_166449.5 | NP_726071.1 | 5 - 6 | 869 | 60 | ||
| NM_166450.3 | NP_726072.2 | 5 - 6 | 1135 | 60 | ||
| GenBank mRNA | AY069377.1 | - | 6 - 7 | 1359 | 60 | |
| AY089610.1 | - | 5 - 6 | 844 | 60 | ||
| BT125826.1 | - | 6 - 7 | 1323 | 60 | ||
More Information
Set Membership: |
|

Sdc
Genomic Map