Assay Details
| Gene Symbol: Fkbp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259519.2 | NP_001246448.1 | 3 - 4 | 426 | 108 | |
| NM_001299750.1 | NP_001286679.1 | 3 - 4 | 421 | 108 | ||
| NM_001299751.1 | NP_001286680.1 | 2 - 3 | 477 | 108 | ||
| NM_057625.5 | NP_476973.1 | 3 - 4 | 426 | 108 | ||
| NM_166452.3 | NP_726074.2 | 3 - 4 | 428 | 108 | ||
| GenBank mRNA | AF132154.2 | - | 3 - 4 | 422 | 108 | |
More Information
Set Membership: |
|

Fkbp14
Genomic Map