Assay Details
| Gene Symbol: Fkbp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299750.1 | NP_001286679.1 | 4 - 5 | 614 | 60 | |
| NM_001299751.1 | NP_001286680.1 | 3 - 4 | 670 | 60 | ||
| NM_057625.5 | NP_476973.1 | 4 - 5 | 619 | 60 | ||
| NM_166452.3 | NP_726074.2 | 4 - 5 | 621 | 60 | ||
| GenBank mRNA | AF132154.2 | - | 4 - 5 | 615 | 60 | |
More Information
Set Membership: |
|

Fkbp14
Genomic Map