Assay Details
| Gene Symbol: blw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166554.3 | NP_726243.1 | 1 - 2 | 167 | 60 | |
| GenBank mRNA | BT003592.1 | - | 1670 | 60 | ||
| Y07894.1 | - | 132 | 60 | |||
More Information
Set Membership: |
|

blw
Genomic Map