Assay Details
| Gene Symbol: RpL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038802.2 | NP_001033891.1 | 3 - 4 | 605 | 81 | |
| NM_001273383.2 | NP_001260312.1 | 2 - 3 | 863 | 81 | ||
| NM_078806.4 | NP_523530.1 | 3 - 4 | 622 | 81 | ||
| GenBank mRNA | BT053725.1 | - | 2 - 3 | 576 | 81 | |
| BT128812.1 | - | 3 - 4 | 607 | 81 | ||
More Information
Set Membership: |
|

RpL13
Genomic Map