Assay Details
Gene Symbol: Prosalpha6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298869.1 | NP_001285798.1 | 2 - 3 | 726 | 66 | |
NM_078808.4 | NP_523532.1 | 2 - 3 | 726 | 66 | ||
GenBank mRNA | BT133448.1 | - | 2 - 3 | 724 | 66 | |
X15497.1 | - | 704 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
