Assay Details
Gene Symbol: Cdk5alpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273391.1 | NP_001260320.1 | 3 - 4 | 1486 | 63 | |
NM_078811.3 | NP_523535.2 | 3 - 4 | 1304 | 63 | ||
GenBank mRNA | AF231134.1 | - | 3 - 4 | 1254 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
