Assay Details
| Gene Symbol: mRpS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273410.1 | NP_001260339.1 | 1 - 2 | 132 | 84 | |
| NM_078813.3 | NP_523537.1 | 1 - 2 | 132 | 84 | ||
| GenBank mRNA | AY075413.1 | - | 1 - 2 | 130 | 84 | |
More Information
Set Membership: |
|

mRpS7
Genomic Map