Assay Details
Gene Symbol: Lip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298882.1 | NP_001285811.1 | 2 - 3 | 565 | 77 | |
NM_078816.4 | NP_523540.1 | 2 - 3 | 565 | 77 | ||
GenBank mRNA | AY075506.1 | - | 2 - 3 | 479 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
