Assay Details
Gene Symbol: Tpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273572.1 | NP_001260501.1 | 1 - 2 | 509 | 75 | |
NM_078860.3 | NP_523584.1 | 1 - 2 | 509 | 75 | ||
GenBank mRNA | AF199022.1 | - | 1 - 2 | 502 | 75 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
