Assay Details
Gene Symbol: Tsp42En | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273829.1 | NP_001260758.1 | 4 - 5 | 776 | 71 | |
NM_078916.3 | NP_523640.1 | 4 - 5 | 776 | 71 | ||
GenBank mRNA | AY071666.1 | - | 4 - 5 | 749 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
