Assay Details
| Gene Symbol: mRpL42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201970.2 | NP_001188899.1 | 1 | 413 | 63 | |
| NM_078949.4 | NP_523673.1 | 1 | 546 | 63 | ||
| GenBank mRNA | AY070620.1 | - | 1 | 547 | 63 | |
More Information
Set Membership: |
|

mRpL42
Genomic Map