Assay Details
| Gene Symbol: Asap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014504.3 | NP_001014504.1 | 7 - 8 | 2662 | 68 | |
| NM_001169614.2 | NP_001163085.1 | 7 - 8 | 2662 | 68 | ||
| NM_001299254.1 | NP_001286183.1 | 7 - 8 | 2662 | 68 | ||
| GenBank mRNA | BT031167.1 | - | 7 - 8 | 2514 | 68 | |
More Information
Set Membership: |
|

Asap
Genomic Map