Assay Details
| Gene Symbol: Asap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014504.3 | NP_001014504.1 | 8 - 9 | 2783 | 76 | |
| GenBank mRNA | BT031167.1 | - | 8 - 9 | 2635 | 76 | |
More Information
Set Membership: |
|

Asap
Genomic Map