Assay Details
| Gene Symbol: Asap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014504.3 | NP_001014504.1 | 10 - 11 | 3705 | 107 | |
| NM_001169614.2 | NP_001163085.1 | 10 - 11 | 3621 | 107 | ||
| GenBank mRNA | BT031167.1 | - | 10 - 11 | 3557 | 107 | |
More Information
Set Membership: |
|

Asap
Genomic Map