Assay Details
Gene Symbol: Cirl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273878.1 | NP_001260807.1 | 3 - 4 | 850 | 98 | |
NM_001273879.2 | NP_001260808.1 | 3 - 4 | 854 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
