Assay Details
Gene Symbol: Cirl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144148.4 | NP_001137620.1 | 2 - 3 | 1818 | 80 | |
NM_001169619.4 | NP_001163090.1 | 2 - 3 | 1818 | 80 | ||
NM_001259275.2 | NP_001246204.1 | 2 - 3 | 1818 | 80 | ||
NM_001259276.2 | NP_001246205.1 | 4 - 5 | 2683 | 80 | ||
NM_001273878.1 | NP_001260807.1 | 5 - 6 | 2509 | 80 | ||
NM_001273879.2 | NP_001260808.1 | 5 - 6 | 2513 | 80 | ||
NM_001273880.2 | NP_001260809.1 | 5 - 6 | 2374 | 80 | ||
NM_136553.5 | NP_610397.4 | 5 - 6 | 2449 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
