Assay Details
| Gene Symbol: Cirl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144148.4 | NP_001137620.1 | 3 - 4 | 2044 | 71 | |
| NM_001259276.2 | NP_001246205.1 | 5 - 6 | 2909 | 71 | ||
| NM_001273878.1 | NP_001260807.1 | 6 - 7 | 2735 | 71 | ||
| NM_136553.5 | NP_610397.4 | 6 - 7 | 2675 | 71 | ||
More Information
Set Membership: |
|

Cirl
Genomic Map