Assay Details
Gene Symbol: Cirl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144148.4 | NP_001137620.1 | 6 - 7 | 2659 | 75 | |
NM_001169619.4 | NP_001163090.1 | 6 - 7 | 2647 | 75 | ||
NM_001259276.2 | NP_001246205.1 | 8 - 9 | 3524 | 75 | ||
NM_001273878.1 | NP_001260807.1 | 9 - 10 | 3350 | 75 | ||
NM_136553.5 | NP_610397.4 | 9 - 10 | 3290 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
