Assay Details
| Gene Symbol: Cirl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144148.4 | NP_001137620.1 | 7 - 8 | 2897 | 64 | |
| NM_001169619.4 | NP_001163090.1 | 7 - 8 | 2885 | 64 | ||
| NM_001259276.2 | NP_001246205.1 | 9 - 10 | 3762 | 64 | ||
| NM_001273878.1 | NP_001260807.1 | 10 - 11 | 3588 | 64 | ||
| NM_136553.5 | NP_610397.4 | 10 - 11 | 3528 | 64 | ||
More Information
Set Membership: |
|

Cirl
Genomic Map