Assay Details
Gene Symbol: Mad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136633.4 | NP_610477.2 | 3 - 4 | 2093 | 73 | |
GenBank mRNA | AF123262.1 | - | 3 - 4 | 2039 | 73 | |
BT044526.1 | - | 3 - 4 | 2083 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
