Assay Details
| Gene Symbol: brp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043070.1 | NP_001036535.1 | 2 - 3 | 585 | 76 | |
| NM_001103771.4 | NP_001097241.2 | 5 - 6 | 1486 | 76 | ||
| NM_001259299.3 | NP_001246228.1 | 5 - 6 | 1486 | 76 | ||
| NM_001259300.2 | NP_001246229.1 | 5 - 6 | 1486 | 76 | ||
| NM_001259301.2 | NP_001246230.1 | 5 - 6 | 1486 | 76 | ||
| NM_001273897.2 | NP_001260826.1 | 4 - 5 | 1095 | 76 | ||
| NM_165677.4 | NP_724796.3 | 6 - 7 | 1730 | 76 | ||
| NM_206063.3 | NP_995785.2 | 4 - 5 | 1095 | 76 | ||
More Information
Set Membership: |
|

brp
Genomic Map